Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Canine distemper virus, Mouse, Clone: 5-4, Abnova™
Mouse monoclonal antibody raised against envelope protein from canine distemper virus.
Supplier: Abnova Corporation MAB0394
Specifications
| Canine distemper virus | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA | |
| 5-4 | |
| Mouse monoclonal antibody raised against envelope protein from canine distemper virus. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Envelope protein from canine distemper virus. | |
| RUO | |
| This antibody is specific for surface envelope antigen of Canine distemper virus. | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction