Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine distemper virus, Mouse, Clone: 8-1, Abnova™

Catalog No. 89134610 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-610 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-610 Supplier Abnova Corporation Supplier No. MAB0393
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against envelope protein from canine distemper virus.

Specifications

Antigen Canine distemper virus
Applications ELISA
Classification Monoclonal
Clone 8-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against envelope protein from canine distemper virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Envelope protein from canine distemper virus.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for surface envelope antigen of Canine distemper virus.
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.