Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPN1, Mouse, Clone: 15C10, Abnova™

Catalog No. 89110656 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-656 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-656 Supplier Abnova Corporation Supplier No. MAB1837
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CAPN1.

The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. [provided by RefSeq

Specifications

Antigen CAPN1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CAPN1.
Dilution Western Blot (0.5-1.0 ug/mL) ELISA (0.5-1.0 ug/mL) Immunoprecipitation (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 150mM NaCl, pH 7.5 (50% glycerol, 3mM sodium azide)
Gene CAPN1
Gene Alias CANP/CANP1/CANPL1/muCANP/muCL
Gene Symbols CAPN1
Host Species Mouse
Immunogen Native purified human CAPN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 823
Test Specificity This antibody recognizes the 80 KDa subunit of micro-calpain as well as two smaller proteins that are presumed to be autolysis products.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.