Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZA1, Mouse, Clone: 2, Abnova™

Catalog No. 89113869 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-869 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-869 Supplier Abnova Corporation Supplier No. MAB9911
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CAPZA1.

CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments. [provided by RefSeq

Specifications

Antigen CAPZA1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CAPZA1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene CAPZA1
Gene Accession No. NM_006135.2
Gene Alias CAPPA1/CAPZ/CAZ1
Gene Symbols CAPZA1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CAPZA1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 829
Test Specificity It can expression in HeLa whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.