Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD11, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107448 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-448 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-448 Supplier Abnova Corporation Supplier No. H00084433A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CARD11.

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. [provided by RefSeq

Sequence: KYFLPYHPPQRRMNLKGIQLQRAKSPISLKRTSDFQAKGHEEEGTDASPSSCGSLPITNSFTKMQPPRSRSSIMSITAEPPGNDSIVRRYKEDAPHRSTV

Specifications

Antigen CARD11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CARD11.
Formulation 50% glycerol
Gene CARD11
Gene Accession No. NM_032415
Gene Alias BIMP3/CARMA1/MGC133069
Gene Symbols CARD11
Host Species Mouse
Immunogen CARD11 (NP_115791, 481 a.a. to 580 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84433
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.