Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP6, Mouse, Clone: MCH2 14 1-190, Abnova™

Catalog No. 89136509 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-509 Supplier Abnova Corporation Supplier No. MAB8425
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CASP6.

Specifications

Antigen CASP6
Applications Western Blot
Classification Monoclonal
Clone MCH2 14 1-190
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CASP6.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Mouse
Immunogen A mixture of recombinant human CASP6 and mouse myeloma cells corresponding to CASP6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.