Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP7, Mouse, Clone: B4-G2, Abnova™

Catalog No. 89113472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-472 Supplier Abnova Corporation Supplier No. MAB8485

Mouse monoclonal antibody raised against synthetic peptide of CASP7.

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The precursor of this caspase is cleaved by caspase 3 and 10. It is activated upon cell death stimuli and induces apoptosis. Alternative splicing results in four transcript variants, encoding three distinct isoforms. [provided by RefSeq

Specifications

Antigen CASP7
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B4-G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CASP7.
Dilution Western Blot (0.5-2 ug/mL) Immunoprecipitation (4-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 1.5mM thimerosal)
Gene CASP7
Gene Alias CMH-1/ICE-LAP3/MCH3
Gene Symbols CASP7
Host Species Mouse
Immunogen A synthetic peptide corresponding to human CASP7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 840
Test Specificity Detects both pro-caspase-7 (34 KDa) and the large subunit of cleaved caspase-7 (19 KDa) in apoptosis cells on SDS-PAGE immunoblots.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.