Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASR, Mouse, Clone: HL 1499, Abnova™

Catalog No. 89110956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-956 Supplier Abnova Corporation Supplier No. MAB2357
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CASR.

The protein encoded by this gene is a G protein-coupled receptor that is expressed in the parathyroid hormone (PTH)-producing chief cells of the parathyroid gland, and the cells lining the kidney tubule. It senses small changes in circulating calcium concentration and couples this information to intracellular signaling pathways that modify PTH secretion or renal cation handling, thus this protein plays an essential role in maintaining mineral ion homeostasis. Mutations in this gene cause familial hypocalciuric hypercalcemia, familial, isolated hypoparathyroidism, and neonatal severe primary hyperparathyroidism. [provided by RefSeq

Specifications

Antigen CASR
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone HL 1499
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CASR.
Dilution ELISA (1:50-1:200) Immunohistochemistry (Frozen sections) (1:50-1:200) RadioEnzymeImmunoassay (1:50-1:200) Western Blot (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene CASR
Gene Alias CAR/FHH/FIH/GPRC2A/HHC/HHC1/MGC138441/NSHPT/PCAR1
Gene Symbols CASR
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus extracellular region of human CASR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 846
Test Specificity This antibody is specific to Calcium Sensing Receptor protein.
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.