Learn More
CBL, Mouse, Clone: M159, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CBL.
Supplier: Abnova Corporation MAB1378
Description
The cbl oncogene was first identified as part of a transforming retrovirus which induces mouse pre-B and pro-B cell lymphomas. As an adaptor protein for receptor protein-tyrosine kinases, it positively regulates receptor protein-tyrosine kinase ubiquitination in a manner dependent upon its variant SH2 and RING finger domains. Ubiquitination of receptor protein-tyrosine kinases terminates signaling by marking active receptors for degradation. [provided by RefSeq
Specifications
CBL | |
Monoclonal | |
Unconjugated | |
ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
CBL | |
CBL | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CBL. | |
RUO | |
867 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
M159 | |
Mouse monoclonal antibody raised against synthetic peptide of CBL. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
C-CBL/CBL2/RNF55 | |
Mouse | |
100 μL | |
Primary | |
This sequence is highly conserved in rat and mouse c-Cbl. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.