Learn More
CCL19, Mouse, Clone: 3E9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CCL19.
Supplier: Abnova Corporation H00006363M03A
Description
This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene may play a role in normal lymphocyte recirculation and homing. It also plays an important role in trafficking of T cells in thymus, and in T cell and B cell migration to secondary lymphoid organs. It specifically binds to chemokine receptor CCR7. [provided by RefSeq
Sequence: MALLLALSLLVLWTSPAPTLSGTNDAEDCCLSVTQKPIPGYIVRNFHYLLIKDGCRVPAVVFTTLRGRQLCAPPDQPWVERIIQRLQRTSAKMKRRSSSpecifications
CCL19 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC027968 | |
CCL19 | |
CCL19 (AAH27968,1 a.a. ∽ 98 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
6363 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA | |
3E9 | |
Mouse monoclonal antibody raised against a full-length recombinant CCL19. | |
CCL19 | |
CKb11/ELC/MGC34433/MIP-3b/MIP3B/SCYA19 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.