Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2, Mouse, Clone: 6h75, Abnova™

Catalog No. 89113611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-611 Supplier Abnova Corporation Supplier No. MAB8839
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CCL2.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4. [provided by RefSeq

Specifications

Antigen CCL2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6h75
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CCL2.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene CCL2
Gene Alias GDCF-2/HC11/HSMCR30/MCAF/MCP-1/MCP1/MGC9434/SCYA2/SMC-CF
Gene Symbols CCL2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human CCL2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity This antibody recognizes recombinant protein CCL2.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.