Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL23, Mouse, Clone: NYRhMIP3, Abnova™

Catalog No. 89113456 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-456 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-456 Supplier Abnova Corporation Supplier No. MAB8384
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CCL23.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist at CC chemokine receptor 1. Two alternatively spliced variants encoding different active isoforms have been identified. [provided by RefSeq

Specifications

Antigen CCL23
Applications ELISA
Classification Monoclonal
Clone NYRhMIP3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human CCL23.
Dilution The optimal working dilution should be determined by the end user.
Formulation No additive
Gene CCL23
Gene Alias CK-BETA-8/CKb8/Ckb-8/Ckb-8-1/MIP-3/MIP3/MPIF-1/SCYA23
Gene Symbols CCL23
Host Species Mouse
Immunogen Recombinant protein corresponding to human CCL23.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6368
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 500μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.