Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL27/CTACK Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $2020.00
Specifications
Antigen | CCL27/CTACK |
---|---|
Clone | 124308 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.5-3.0 ug/mL |
Applications | Western Blot, Immunohistochemistry, Neutralization |
Classification | Monoclonal |
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CCL27/CTACK | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.5-3.0 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
NP_006655 | |
10850 | |
E. coli-derived recombinant human CCL27/CTACK Phe25-Gly112 Accession # NP_006655 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCL27/CTACK in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 9/10, 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20 is observed. |
124308 | |
Western Blot, Immunohistochemistry, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
CCL27 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title