Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3, Mouse, Clone: 103, Abnova™

Catalog No. 89112814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-814 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-814 Supplier Abnova Corporation Supplier No. MAB6303
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CCL3.

Macrophage inflammatory protein-1 is a so-called monokine that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes (Wolpe et al., 1988 [PubMed 3279154]). Sherry et al. (1988) [PubMed 3058856] demonstrated 2 protein components of MIP1, called by them alpha and beta.[supplied by OMIM

Specifications

Antigen CCL3
Applications ELISA
Classification Monoclonal
Clone 103
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CCL3.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene CCL3
Gene Alias G0S19-1/LD78ALPHA/MIP-1-alpha/MIP1A/SCYA3
Gene Symbols CCL3
Host Species Mouse
Immunogen Recombinant protein corresponding to human CCL3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity This antibody is reactive with recombinant human CCL3.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.