Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCM2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CCM2.
Supplier: Abnova Corporation H00083605A01
Description
Sequence: EEEGKKGKKPGIVSPFKRVFLKGEKSRDKKAHEKVTERRPLHTVVLSLPERVEPDRLLSDYIEKEVKYLGQLTSIPGYLNPSSRTEILHFIDNAKRAHQSpecifications
CCM2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CCM2. | |
CCM2 | |
C7orf22/MGC4067/MGC4607/MGC74868/PP10187 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_031443 | |
CCM2 | |
CCM2 (NP_113631, 2 a.a. to 100 a.a) partial recombinant protein with GST tag. | |
RUO | |
83605 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction