Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNB1, Mouse, Clone: V92.1, Abnova™

Catalog No. 89109532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-532 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-532 Supplier Abnova Corporation Supplier No. MAB11371
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CCNB1.

The protein encoded by this gene is a regulatory protein involved in mitosis. The gene product complexes with p34(cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. The different transcripts result from the use of alternate transcription initiation sites. [provided by RefSeq

Specifications

Antigen CCNB1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone V92.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CCNB1.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (0.5-1 ug/mL) The optimal working dilution s
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene CCNB1
Gene Alias CCNB
Gene Symbols CCNB1
Host Species Mouse
Immunogen Hamster CCNB1.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 891
Target Species Hamster, Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.