Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ccnb2, Mouse, Clone: X29.2, Abnova™

Catalog No. 89113109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-109 Supplier Abnova Corporation Supplier No. MAB6990
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against ccnb2.

Specifications

Antigen Ccnb2
Applications Western Blot
Classification Monoclonal
Clone X29.2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against ccnb2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene LOC397743
Gene Symbols LOC397743
Host Species Mouse
Immunogen Xenopus ccnb2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397743
Test Specificity Reacts with most cyclin Bs. Western Blot Predicted molecular weight 45 KDa.
Target Species Human, Mouse, Rat, Xenopus species
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.