Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNB3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107482 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-482 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-482 Supplier Abnova Corporation Supplier No. H00085417A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CCNB3.

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. Studies of similar genes in chick and Drosophila suggest that this cyclin may associate with CDC2 and CDK2 kinases, and be required for proper spindle reorganization and restoration of the interphase nucleus. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: VQEKASKLAAASLLLALYMKKLGYWVPFLEHYSGYSISELHPLVRQLNKLLTFSSYDSLKAVYYKYSHPVFFEVAKIPALDMLKLEEILNCDCEAQGLVL

Specifications

Antigen CCNB3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CCNB3.
Formulation 50% glycerol
Gene CCNB3
Gene Accession No. NM_033031
Gene Symbols CCNB3
Host Species Mouse
Immunogen CCNB3 (NP_149020, 1296 a.a. to 1395 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85417
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.