Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCND1, Mouse, Clone: CD1.1, Abnova™

Catalog No. 89112908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-908 Supplier Abnova Corporation Supplier No. MAB6501
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CCND1.

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance throughout the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK4 or CDK6, whose activity is required for cell cycle G1/S transition. This protein has been shown to interact with tumor suppressor protein Rb and the expression of this gene is regulated positively by Rb. Mutations, amplification and overexpression of this gene, which alters cell cycle progression, are observed frequently in a variety of tumors and may contribute to tumorigenesis. [provided by RefSeq

Specifications

Antigen CCND1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CD1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CCND1.
Dilution Flow Cytometry Immunocytochemistry (1 ug/mL) Immunohistochemistry (2 ug/mL) Immunoprecipitation (1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CCND1
Gene Alias BCL1/D11S287E/PRAD1/U21B31
Gene Symbols CCND1
Host Species Mouse
Immunogen Purified cyclin D1 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 595
Test Specificity This antibody recognizes CCND1, an ubiquitously expressed 33 KDa protein thatmigrates as a 36 KDa band under reducing SDS-PAGE conditions.
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.