Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCND1, Mouse, Clone: CY-D1, Abnova™

Catalog No. 89110436 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-436 Supplier Abnova Corporation Supplier No. MAB1449
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CCND1.

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance throughout the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK4 or CDK6, whose activity is required for cell cycle G1/S transition. This protein has been shown to interact with tumor suppressor protein Rb and the expression of this gene is regulated positively by Rb. Mutations, amplification and overexpression of this gene, which alters cell cycle progression, are observed frequently in a variety of tumors and may contribute to tumorigenesis. [provided by RefSeq

Specifications

Antigen CCND1
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone DCS-6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CCND1.
Dilution Western Blot (4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (8 ug/mL) Immunohistochemistry (Frozen sections) (8 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene CCND1
Gene Alias BCL1/D11S287E/PRAD1/U21B31
Gene Symbols CCND1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CCND1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 595
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.