Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1, Mouse, Clone: 76-7-4, Abnova™
Mouse monoclonal antibody raised against CD1.
Supplier: Abnova Corporation MAB6216
Specifications
| CD1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD1 | |
| Mouse | |
| 500 μg | |
| Primary | |
| Porcine CD1, Mr 43-49 KDa. | |
| Store at 4°C. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| 76-7-4 | |
| Mouse monoclonal antibody raised against CD1. | |
| In 100mM BBS, pH 8.2 | |
| CD1 | |
| dd Miniature swine thymocytes. | |
| RUO | |
| 396800 | |
| Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction