Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1, Mouse, Clone: RIV12, Abnova™

Catalog No. 89112165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-165 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-165 Supplier Abnova Corporation Supplier No. MAB5204
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD1.

Specifications

Antigen CD1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RIV12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Alias CD1/FCB6/HTA1/R4/T6
Gene Symbols CD1A
Host Species Mouse
Immunogen Native purified CD1 from human peripheral lymphocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 909/910/911/912/913
Test Specificity RIV12, selected by ELISA, cytotoxicity tests and FACS analysis appeared to be an /CD1 reacting with a 49 KDa molecule on human thymocytes. Splenocytes of the mouse were fused with mouse myeloma cells of the 653 cell line.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.