Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107b Mouse anti-Human, BUV395, Clone: H4B4, BD

Catalog No. BDB565303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565303 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565303 Supplier BD Biosciences Supplier No. 565303

Mouse Monoclonal Antibody

The H4B4 monoclonal antibody specifically binds to CD107b. CD107b is a heavily glycosylated, ∽120 kDa type I transmembrane protein that is also known as lysosomal-associated membrane protein 2 (LAMP-2/LAMP2), LAMPB, or LGP110. CD107b is expressed in the lysosomal and endosomal membranes of most cell types where it may play a role in maintaining membrane integrity. CD107b is also expressed on the surface of activated platelets and lymphocytes, including T lymphocytes, e.g., degranulating CD8+ cytotoxic T cells. CD107b serves as a ligand for selectins, including E-selectin, and mediates cellular adhesion. CD107a/ LAMP-1 and CD107b /LAMP-2 are carriers for poly-N-acetyllactosamines, and display the sialyl Le[x] antigen. CD107b is likewise expressed on some tumor cells and cell lines, including U937 and KG1a, and may facilitate tumor metastases.

The antibody was conjugated to BD Horizon BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107b
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone H4B4
Conjugate Brilliant Ultraviolet 395
Description LAMP-2; LAMP2; LAMPB; LGP110
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Adult Adherent Peripheral Blood Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.