Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Mouse anti-Human, Brilliant Violet 421, Clone: 3.9, BD

Catalog No. BDB565807 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565807 25 μg
BDB565806 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565807 Supplier BD Biosciences Supplier No. 565807
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3.9 monoclonal antibody specifically binds to CD11c, which is also known as Integrin alpha X ( α X Integrin/ITGAX), or p150,95 Integrin alpha chain. CD11c is a ∽150 kDa type I transmembrane glycoprotein. It is expressed on monocytes, macrophages, granulocytes, NK cells, dendritic cells, and subsets of B and T cells. It associates with CD18 (Integrin beta 2/ β 2 Integrin) to form the CD11c/CD18 complex, which is also known as p150,95 Integrin, or the Type 4 Complement Receptor (CR4). CD11c/CD18 binds fibrinogen and reportedly serves as a receptor for iC3b and ICAM-1/CD54. CD11c/CD18 functions as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium. The 3.9 monoclonal antibody crossreacts with Rhesus macaque leucocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 3.9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description ITGAX; AlphaX Integrin; Axb2; Integrin alpha-X; CR4; SLEB6; p150,95 alpha
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human monocytes and synovial cells
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.