Learn More
CD276 Mouse anti-Human, Unlabeled, Clone: 7-517, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565828
Description
The 7-517 monoclonal antibody specifically binds to CD276, also known as B7-H3 (B7 homolog 3). CD276 is a type I transmembrane glycoprotein and member of the B7 family of immunoregulatory proteins. Although the CD276 gene transcript is expressed in nearly all normal tissues, the protein is preferentially expressed in tumors, tumor cell lines, sinonasal epithelial cells, and extravillous trophoblast and Hofbauer cells of the placenta. CD276 protein expression can be induced by activation of T and B lymphocytes, natural killer (NK) cells and antigen-presenting cells. The roles of CD276 in the regulation of immune responses, especially to cancer, are under investigation.

Specifications
CD276 | |
Monoclonal | |
0.5mg/mL | |
B7-H3, B7H3, 4Ig-B7-H3, B7RP-2 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
7-517 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Monocyte-Derived Dendritic Cells | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.