Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14, Mouse, Clone: biG 13, Abnova™

Catalog No. 89112345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-345 Supplier Abnova Corporation Supplier No. MAB5654
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD14.

CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq

Specifications

Antigen CD14
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone biG 13
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CD14.
Dilution Flow Cytometry (1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene CD14
Gene Symbols CD14
Host Species Mouse
Immunogen Monocytes of different species and immunoaffinity purified soluble human CD14.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Test Specificity biG 13 represents an excellent marker for CD14. CD14 has been shown to be involved in LPS-induced cell activation. This antigen is present as a membrane bound form and as a soluble antigen, both of which are involved in cell stimulation by LPS. The plasma protein LBP plays an important role in the LPS-CD14 mediated cell activation. The CD14 glycoprotein, gp 55, is present on most monocytic and macrophages like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for Endotoxin.
Target Species Bovine, Canine, Equine, Human, Pig
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.