Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14, Mouse, Clone: biG 53, Abnova™

Catalog No. 89112351 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-351 Supplier Abnova Corporation Supplier No. MAB5660
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Cd14.

Specifications

Antigen CD14
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone biG 53
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Cd14.
Dilution ELISA (1:20000) Flow Cytometry (1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Cd14
Gene Symbols Cd14
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Cd14.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12475
Target Species Mouse
Content And Storage Store at -80°C.
After reconstitution with 100 uL of deionized water and 0.1% BSA, store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.