Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-CD14, Clone: MDA/929, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | CD14 |
---|---|
Clone | MDA/929 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24486602
![]() |
Novus Biologicals
NBP2448660.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
CD14 | |
0.2 mg/mL | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
929 | |
Recombinant human CD14 protein | |
Primary | |
Store at 4C. | |
55 kDa |
MDA/929 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
CD14 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 55kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title