Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14, Mouse, Clone: UCHM-1, Abnova™

Catalog No. 89135622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-622 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-622 Supplier Abnova Corporation Supplier No. MAB4129

Mouse monoclonal antibody raised against native CD14.

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone UCHM-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD14.
Dilution Flow Cytometry (1ug stain 1.0x106 cells) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified CD14 from thymocytes and peripheral blood leucocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a (Mr 55 KDa) cell surface glycoprotein expressed on most peripheral blood monocytes. CD14 antigen is expressed on 70-95% of peripheral blood monocytes. It is also found on other cells of myeloid lineage including granulocytes, dendritic reticulum cells and pleural phagocytic cells.
Content And Storage Store at 4°C.
Product Type CD14 monoclonal antibody, clone UCHM-1
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.