Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16, Mouse, Clone: eBioCB16, Abnova™

Catalog No. 89109083 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-083 Supplier Abnova Corporation Supplier No. MAB10877

Mouse monoclonal antibody raised against CD16.

Specifications

Antigen CD16
Applications Flow Cytometry
Classification Monoclonal
Clone eBioCB16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CD16.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Alias CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3
Gene Symbols FCGR3A
Host Species Mouse
Immunogen Human CD16.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214/2215
Test Specificity The monoclonal antibody recognizes CD16 (Fc gamma RIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80 kDa.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.