Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD175, Mouse, Clone: BRIC111, Abnova™

Catalog No. 89135923 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-923 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-923 Supplier Abnova Corporation Supplier No. MAB5231
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD175.

Specifications

Antigen CD175
Applications Agglutination, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BRIC111
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD175.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified CD175 from human Tn red blood cells.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes Tn antigen on glycophorin A and glycophorin B in human erythrocytes. Tn is a cryptantigen which was recently designated CD175 at the 7th Leucocyte Typing Workshop. Tn antigen is not expressed on normal haemopoietic cells but exposure of the Tn is associated with polyagglutination.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.