Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18, Mouse, Clone: IVA35, Abnova™

Catalog No. 89108308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-308 Supplier Abnova Corporation Supplier No. MAB0986
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD18.

Specifications

Antigen CD18
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone IVA35
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD18.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ITGB2
Gene Symbols ITGB2
Host Species Mouse
Immunogen Native purified CD18 from bovine leukocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281877
Target Species Bovine
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.