Learn More
CD180, Mouse, Clone: G28-8, Abnova™
Mouse monoclonal antibody raised against CD180.
Supplier: Abnova Corporation MAB10171
Description
CD180 is a cell surface molecule consisting of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is associated with a molecule called MD-1 and form the cell surface receptor complex, RP105/MD-1. It belongs to the family of pathogen receptors, Toll-like receptors (TLR). RP105/MD1, by working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS), a membrane constituent of Gram-negative bacteria. [provided by RefSeq
Specifications
CD180 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (2 ug/mL) The optimal working dilution should be determined by the end user. | |
CD180 | |
CD180 | |
Human tonsillar B cells. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Functional Assay, Immunoprecipitation | |
G28-8 | |
Mouse monoclonal antibody raised against CD180. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
LY64/Ly78/MGC126233/MGC126234/RP105 | |
Mouse | |
Protein A purification | |
RUO | |
4064 | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.