Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd19, Mouse, Clone: MB19-1, Abnova™

Catalog No. 89111275 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-275 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-275 Supplier Abnova Corporation Supplier No. MAB3227
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cd19.

Specifications

Antigen Cd19
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MB19-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cd19.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Cd19
Gene Alias AW495831
Gene Symbols Cd19
Host Species Mouse
Immunogen 300.191., CD19+ mouse pre-B cell line
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12478
Test Specificity CD19 B-cell differentiation antigen, Mr 95 KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.