Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19, Mouse, PE, Clone: 4G7, Abnova™

Catalog No. 89108439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-439 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-439 Supplier Abnova Corporation Supplier No. MAB10234
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of CD19.

Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. [provided by RefSeq

Specifications

Antigen CD19
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 4G7
Conjugate PE
Description Mouse monoclonal antibody raised against of CD19.
Dilution Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD19
Gene Alias B4/MGC12802
Gene Symbols CD19
Host Species Mouse
Immunogen Human CCL (chronic lymphocytic leukemia) cells.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 930
Test Specificity CD19, type I transmembrane glycoprotein of immunoglobulin superfamily, expressed on B lymphocytes and follicular dendritic cells; it is lost on plasma cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.