Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-CD1b, Clone: 100-1A5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452430.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD1b Monoclonal antibody specifically detects CD1b in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
| CD1b | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| P29016 | |
| CD1B | |
| Stimulated human leukocytes | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The mouse monoclonal antibody recognizes CD1b, a 44kDa type I glycoprotein associated with beta2-microglobulin (Workshop IV; Code T015). It is expressed on dendritic cells, Langerhans cells, thymocytes, and T acute lymphoblastic leukemia cells. The CD1 multigene family encodes five forms of the CD1 T-cell surface glycoprotein in human, designated CD1A, 1B, 1C, 1D and 1E. CD1, a type 1 membrane protein, has structural similarity to the MHC class I antigen and has been shown to present lipid antigens for recognition by T lymphocytes. Constitutive endocytosis of CD1B molecules and the differential sorting of MHC class II from lysosomes separate peptide- and lipid antigen-presenting molecules during dendritic cell maturation. CD1B is also expressed in interdigitating cells. | |
| Store at 4C. | |
| IgM κ |
| Flow Cytometry, Immunofluorescence | |
| 100-1A5 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
| Mouse | |
| 44 kDa | |
| 0.2 mg | |
| Immunology | |
| 910 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction