Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CD1d, Clone: D5,

Catalog No. MABC983MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC983MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC983MI Supplier MilliporeSigma™ Supplier No. MABC983
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD1d clone: D5 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

Antigen-presenting glycoprotein CD1d (UniProt P15813; also known as CD1d, R3G1) is encoded by the CD1D gene (Gene ID 912) in human. CD1d is an MHC-like glycoprotein expressed on epithelial cells and can signal innate immune responses by presenting lipid antigen to natural killer (NK) and NK T (NKT) cells. Similar to MHC class I heavy chain (HC), CD1d HC is glycosylated and assembled with β2m in ER. Unlike MHC class I HC, misfoled CD1d and CD1d not properly assembled with β2m are not destroyed by ER quality control mechanisms. CD1d plays a role in both innate and adaptive immunity to various bacteria, viruses, fungi, and parasites. Activation of CD1d-restricted invariant NKT cells enhances host resistance to these microbes via direct killing of CD1d-expressing (infected) cells or by promoting interferon gamma release from conventional NK cells and by modulating adaptive immune cells via alteration of Th1/Th2 polarization. Chlamydia trachomatis is shown to prevent CD1d-mediated innate immune destruction of the infected urethral epithelial cells by targeting CD1d for proteolytic degradation. Specifically, the chlamydial proteasome-like activity factor (CPAF) interacts with the CD1d HC, and CPAF-associated CD1d HC is then ubiquitinated for eventual degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1d
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D5
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P15813
Gene Symbols CD1D
Host Species Mouse
Immunogen Pooled CD1d-GST fusion proteins containing α1 domain alone, α1-α2 domains, or α1–α3 domains.
Purification Method Protein G Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001757
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.