Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2, Mouse, Biotin, Clone: MEM-65, Abnova™

Catalog No. 89111896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-896 Supplier Abnova Corporation Supplier No. MAB4556
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD2.

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-65
Conjugate Biotin
Description Mouse monoclonal antibody raised against native CD2.
Dilution Flow Cytometry (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene CD2
Gene Alias SRBC/T11
Gene Symbols CD2
Host Species Mouse
Immunogen Native purified CD2 from human peripheral T cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914
Test Specificity This antibody recognizes an unique epitope of CD2, a 50 KDa glycoprotein present on the human peripheral blood T-lymphocytes and NK cells; also expressed by all thymocytes.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.