Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd2, Mouse, Clone: MRC OX-34, Abnova™

Catalog No. 89109318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-318 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-318 Supplier Abnova Corporation Supplier No. MAB11136

Mouse monoclonal antibody raised against Cd2.

Specifications

Antigen Cd2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Monoclonal
Clone MRC OX-34
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cd2.
Dilution Flow Cytometry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Cd2
Gene Alias CD2R/OX-34
Gene Symbols Cd2
Host Species Mouse
Immunogen Rat Cd2 from activated rat T helper cells.
Purification Method Protein G purification
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497761
Target Species Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.