Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD22, Mouse, Clone: FPC1, Abnova™

Catalog No. 89112190 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-190 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-190 Supplier Abnova Corporation Supplier No. MAB5253

Mouse monoclonal antibody raised against recombinant CD22.

Specifications

Antigen CD22
Applications Immunohistochemistry
Classification Monoclonal
Clone FPC1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CD22.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:40) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Gene CD22
Gene Alias FLJ22814/MGC130020/SIGLEC-2/SIGLEC2
Gene Symbols CD22
Host Species Mouse
Immunogen Recombinant protein corresponding to human CD22.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 933
Test Specificity This antibody recognizes the human CD22 cell surface antigen, a 130-140kD glycoprotein.CD22 is expressed within the cytoplasm at an early stage of B cell differentiation, but is expressed at the membrane only on mature B lymphocytes. CD22 expression is lost during terminal differentiation to plasma cell stages.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.