Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD247, Mouse, Clone: 4D10A6, Abnova™

Catalog No. 89108649 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-649 Supplier Abnova Corporation Supplier No. MAB10463
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CD247.

The protein encoded by this gene is T-cell receptor zeta, which together with T-cell receptor alpha/beta and gamma/delta heterodimers, and with CD3-gamma, -delta and -epsilon, forms the T-cell receptor-CD3 complex. The zeta chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CD247
Applications ELISA, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 4D10A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CD247.
Dilution Immunohistochemistry (1:200-1:400) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene CD247
Gene Alias CD3-ZETA/CD3H/CD3Q/CD3Z/T3Z/TCRZ
Gene Symbols CD247
Host Species Mouse
Immunogen Recombinant protein corresponding to human CD247.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 919
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.