Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25 Mouse anti-Rat, Brilliant Violet 421, Clone: OX-39, BD

Catalog No. BDB565608 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565608 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565608 Supplier BD Biosciences Supplier No. 565608
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-39 monoclonal antibody specifically b inds to the α chain of the IL-2 R eceptor (IL-2R α , IL2RA) on activated T cells, T regulatory cells, and thymic and splenic dendritic cells. CD25 has also been detected on rat intestinal epithelial cells. It has been reported that OX-39 mAb weakly blocks binding of IL-2 to T-cell blasts and that it blocks IL-2 stimulated epithelial cell migration in an in vitro model of wound healing.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25
Applications Flow Cytometry
Classification Monoclonal
Clone OX-39
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat T blasts from mixed-lymphocyte reactions
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.