Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27, Mouse, FITC, Clone: LT27, Abnova™

Catalog No. 89111804 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-804 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-804 Supplier Abnova Corporation Supplier No. MAB4399
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD27.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-kappaB and MAPK8/JNK. Adaptor proteins TRAF2 and TRAF5 have been shown to mediate the signaling process of this receptor. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor. [provided by RefSeq

Specifications

Antigen CD27
Applications Flow Cytometry
Classification Monoclonal
Clone LT27
Conjugate FITC
Description Mouse monoclonal antibody raised against native CD27.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD27
Gene Alias MGC20393/S152/T14/TNFRSF7/Tp55
Gene Symbols CD27
Host Species Mouse
Immunogen Native purified CD27 from human peripheral blood lymphocytes.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 939
Test Specificity This antibody reacts with CD27 (T14), a 50-55 KDa type I transmembrane glycoprotein (member of the TNF-receptor superfamily) expressed on medullary thymocytes, peripheral T lymphocytes, some B lymphocytes and NK cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.