Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Mouse, Unlabeled, Clone: U8-424, BD

Catalog No. BDB565830 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565830 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565830 Supplier BD Biosciences Supplier No. 565830
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The U8-424 monoclonal antibody specifically binds to human, mouse, and rat C-Peptide, the connecting peptide that links the A- and B-chains in the proinsulin molecule. The A- and B- chains and C-Peptide are encoded by the transcript of the INS gene and are produced by the β cells in the islets of Langerhans of the pancreas . As the biosynthesis of insulin proceeds, the C-Peptide is cleaved from proinsulin to form the mature insulin hormone, which is composed of the A- and B-chains linked by 2 disulfide bonds. Mature insulin and C-Peptide are stored i n granules in the β cells and are released to the blood in response to metabolic signals such as glucose, the amino acids arginine and leucine, and acetylcholine. As a result, C-Peptide is released into the blood stream in an equimolar amount to insulin; the serum level of C- Peptide correlates with pancreatic β cell function and the amount of insulin being produced. The expression of C-Peptide can be used to monitor the pancreatic differentiation of pluripotent stem cells. Insulin is an evolutionarily conserved peptide hormone that binds to receptors on target cells (primarily adipose and muscle) to promote the absorption of glucose from the blood, thus regulating fat and carbohydrate metabolism. C-Peptide itself binds to many cell types, independently of the insulin receptor, and initiates several intracellular signaling cascades.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Peptide
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone U8-424
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human C-Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.