Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD278 Mouse anti-Human, BUV395, Clone: DX29, BD

Catalog No. BDB564777 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564777 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564777 Supplier BD Biosciences Supplier No. 564777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DX29 monoclonal antibody specifically binds to human CD278, which is also known as Inducible Costimulator (ICOS) or Inducible T-cell Costimulator. ICOS is a homodimeric type I transmembrane glycoprotein with an approximate molecular weight of 50-60 kDa. It is a member of the CD28 family and is highly expressed on activated T cells. CD278 is the receptor for ICOS-ligand (also known as, CD275, B7-H2, B7RP-1, or LICOS). Like CD28, ICOS can provide a costimulatory signal for T cell activation, proliferation and cytokine production. It is not expressed on resting or activated B cells, monocytes, NK cells, granulocytes, dendritic cells or platelets. Unlike the constitutively expressed CD28, ICOS is de novo expressed upon cellular activation. Reports describe similarities between CD28 and ICOS in T cell activation, such as the costimulation of cytokine production. However, it has been suggested that ICOS may play a greater role in IL-10 production. In the presence of IL-10, purified recombinant human ICOS protein significantly increased in vitro B cell growth stimulated by pokeweed mitogen (PWM) and enhanced production of IgG.

The antibody was conjugated to BD Horizon BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD278
Applications Flow Cytometry
Classification Monoclonal
Clone DX29
Conjugate Brilliant Ultraviolet 395
Description ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Activated human T cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.