Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-CD28, Clone: CB28, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | CD28 |
---|---|
Clone | CB28 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Frozen 0.5-1ug/ml, Functional, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23459202
![]() |
Novus Biologicals
NBP2345920.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready.Specifications
CD28 | |
1.0 mg/mL | |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Immunology | |
10mM PBS - BSA free with No Preservative | |
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
CD28 | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CB28 | |
Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Frozen 0.5-1ug/ml, Functional, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P10747 | |
940 | |
Recombinant human CD28 protein | |
Primary | |
Protein A purified | |
Recognizes a glycoprotein of 44-88kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title