Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd28, Mouse, Clone: JJ319, Abnova™

Catalog No. 89108423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-423 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-423 Supplier Abnova Corporation Supplier No. MAB10176
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cd28.

Specifications

Antigen Cd28
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone JJ319
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cd28.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Cd28
Gene Alias CD28RNA
Gene Symbols Cd28
Host Species Mouse
Immunogen Mouse A20J B lymphoma cells transfected with rat Cd28.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25660
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.