Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-CD2AP, Clone: 2A2.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABT419
Description
Specifically detects CD2AP clone: 2A2.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
CD2-associated protein (UniProt Q9Y5K6; also known as Adapter protein CMS, Cas ligand with multiple SH3 domains) is encoded by the CD2AP (also known as CMS) gene (Gene ID 23607) in human. CD2AP is an adapter protein expressed in all tissues except brain. CD2AP harbors three SH3 domains at its N-terminal, a proline-rich domain, and a C-terminal leucine-zipper domain that mediates CD2AP homodimerization. CD2AP is known to interact with actin-associated proteins, such as cortactin, and actin capping protein CAPZ. The SH3 domains of CD2AP mediate its association to the hypervariable C terminal region of Rac1, allowing CD2AP to act as a linker between Rac1 and the actin-binding proteins cortactin and CAPZ. CD2AP also plays an important role in the formation of the slit diaphragm in the kidney glomerulus between podocyte foot processes by linking the cell-cell adhesion protein nephrin to the cytoskeleton. CD2AP gene mutations are associated with kidney diseases such as sporadic nephrotic syndrome and focal segmental glomerulosclerosis.
Specifications
CD2AP | |
Monoclonal | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
CD2AP | |
GST-tagged recombinant protein corresponding to the C-terminal half of human CD2AP. | |
100 μg | |
Cell Structure | |
NP_036252 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
2A2.1 | |
Q9Y5K6 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction