Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3, Mouse, Cy5, Clone: UCHT1, Abnova™

Catalog No. 89112618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-618 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-618 Supplier Abnova Corporation Supplier No. MAB6007
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD3.

Specifications

Antigen CD3
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone UCHT1
Conjugate Cyanine5
Description Mouse monoclonal antibody raised against native CD3.
Dilution Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias CD3-DELTA/T3D
Gene Symbols CD3D
Host Species Mouse
Immunogen Native purified from human thymocytes and peripheral blood lymphocytes from a patient with Sezary cell leukemia.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 915/916/917/919
Test Specificity Specificity epsilon chain of the CD3/T cell antigen receptor complex
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.