Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3, Mouse, PE, Clone: APA1/1, Abnova™

Catalog No. 89136782 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-782 Supplier Abnova Corporation Supplier No. MAB9820
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD3.

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone APA1/1
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against native CD3.
Dilution Flow Cytometry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified CD3 from human thymus.
Purification Method Size-exclusion chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The mouse monoclonal antibody APA1/1 recognizes an activation-dependent intracellular epitope of CD3 epsilon. Exposure of the epitope precedes CD3 phosphorylation and recruitment and activation of ZAP70, which initiates the signaling cascade produced by T-cell activation. APA1/1 provides the earliest known marker for TCR-mediated T cell activation.
Target Species Human, Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.